missing translation for 'onlineSavingsMsg'
Learn More

Phthalonitrile 99.0+%, TCI America™

Catalog No. P040425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P040425G 25 g
P0404100G 100 g
P0404500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P040425G Supplier TCI America Supplier No. P040425G

Chemical Identifiers

CAS 91-15-6
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001771
InChI Key XQZYPMVTSDWCCE-UHFFFAOYSA-N
Synonym phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
PubChem CID 7042
IUPAC Name benzene-1,2-dicarbonitrile
SMILES N#CC1=CC=CC=C1C#N

Specifications

Melting Point 140°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 128.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phthalonitrile
EINECSNumber (3)-1799
RTECSNumber TI8575000
TSCA Yes