missing translation for 'onlineSavingsMsg'
Learn More

Phthalonitrile, 98%

CAS: 91-15-6 | C8H4N2 | 128.13 g/mol

Supplier:  Thermo Scientific Chemicals A150710B

 View more versions of this product

Catalog No. AAA150710B


Only null left
Explore available promotions

Description

Description

Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

Solubility
Soluble in alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Phthalonitrile
139°C to 141°C
C8H4N2
1000 g
775028
Soluble in alcohol and ether.
N#CC1=CC=CC=C1C#N
128.13
128.13
91-15-6
162°C (323°F)
MFCD00001771
UN3439
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
XQZYPMVTSDWCCE-UHFFFAOYSA-N
benzene-1,2-dicarbonitrile
7042
98%

Chemical Identifiers

91-15-6
128.13
XQZYPMVTSDWCCE-UHFFFAOYSA-N
7042
N#CC1=CC=CC=C1C#N
C8H4N2
MFCD00001771
phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile
benzene-1,2-dicarbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331-H500

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 202-044-8

RTECSNumber : TI8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only