missing translation for 'onlineSavingsMsg'
Learn More

Phthalocyanine Chloroaluminum 98.0+%, TCI America™

Catalog No. c11671g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
C11671G 1 g
1 options

Catalog No. C11671G

Supplier: TCI America C11671G

Chemical Identifiers

CAS 14154-42-8
Molecular Formula C32H16AlClN8
MDL Number MFCD00049386
Synonym Chloroaluminum Phthalocyanine

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 574.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalocyanine Chloroaluminum