missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide 98.0+%, TCI America™

Catalog No. p-7134588
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P040225G 25 g
P0402500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P040225G Supplier TCI America Supplier No. P040225G
Only null left

Chemical Identifiers

CAS 85-41-6
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00005881
InChI Key XKJCHHZQLQNZHY-UHFFFAOYSA-N
Synonym phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
PubChem CID 6809
ChEBI CHEBI:38817
IUPAC Name 2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1NC(=O)C2=CC=CC=C12

Specifications

Melting Point 234°C
Color White
Quantity 25 g
Formula Weight 147.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalimide
EINECSNumber (5)-0079
RTECSNumber TI3920000
TSCA Yes