missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide Potassium Salt 98.0+%, TCI America™

Catalog No. P0403500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0403500G 500 g
P040325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0403500G Supplier TCI America Supplier No. P0403500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1074-82-4
Molecular Formula C8H4KNO2
Molecular Weight (g/mol) 185.22
MDL Number MFCD00005887
InChI Key FYRHIOVKTDQVFC-UHFFFAOYSA-M
Synonym potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
PubChem CID 3356745
IUPAC Name potassium 1,3-dioxo-2,3-dihydro-1H-isoindol-2-ide
SMILES [K+].O=C1[N-]C(=O)C2=CC=CC=C12

Specifications

Color White
Quantity 500 g
Formula Weight 185.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalimide Potassium Salt
EINECSNumber (5)-5493
TSCA Yes