missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, potassium derivative, 99%

Catalog No. AC170861000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170861000 100 g Plastic bottle
AC170865000 500 g Plastic bottle
AC170860025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC170861000 Supplier Thermo Scientific Chemicals Supplier No. 170861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1074-82-4 | C8H4KNO2 | 185.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1074-82-4
Molecular Formula C8H4KNO2
Molecular Weight (g/mol) 185.22
MDL Number MFCD00005887
InChI Key FYRHIOVKTDQVFC-UHFFFAOYSA-M
Synonym potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
PubChem CID 3356745
IUPAC Name potassium;isoindol-2-ide-1,3-dione
SMILES [K+].O=C1[N-]C(=O)C2=CC=CC=C12

Specifications

Melting Point >300°C
Color Green or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Solubility Information Solubility in water: soluble in water
Formula Weight 185.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Phthalimide, potassium derivative
Show More Show Less
EINECSNumber 214-046-6

RUO – Research Use Only