missing translation for 'onlineSavingsMsg'
Learn More

Phthalide 98.0+%, TCI America™

Catalog No. p-7134587
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P040125G 25 g
P0401500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P040125G Supplier TCI America Supplier No. P040125G
Only null left

Chemical Identifiers

CAS 87-41-2
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00005906
InChI Key WNZQDUSMALZDQF-UHFFFAOYSA-N
Synonym phthalide, isobenzofuran-1 3h-one, 1 3h-isobenzofuranone, 1-phthalanone, phthalolactone, 2-benzofuran-1 3h-one, isobenzofuranone, phthalanone, 3h-isobenzofuran-1-one, 2-hydroxymethylbenzoic acid, gamma-lactone
PubChem CID 6885
ChEBI CHEBI:38085
IUPAC Name 3H-2-benzofuran-1-one
SMILES C1C2=CC=CC=C2C(=O)O1

Specifications

Melting Point 75°C
Color White
Quantity 25 g
Formula Weight 134.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phthalide
EINECSNumber (5)-0018
RTECSNumber TI3520000
TSCA Yes