missing translation for 'onlineSavingsMsg'
Learn More

Phthalide, 98+%

Catalog No. AAA1551622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1551622 100 g
AAA1551636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1551622 Supplier Thermo Scientific Chemicals Supplier No. A1551622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-41-2 | C8H6O2 | 134.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-41-2
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00005906
InChI Key WNZQDUSMALZDQF-UHFFFAOYSA-N
Synonym phthalide, isobenzofuran-1 3h-one, 1 3h-isobenzofuranone, 1-phthalanone, phthalolactone, 2-benzofuran-1 3h-one, isobenzofuranone, phthalanone, 3h-isobenzofuran-1-one, 2-hydroxymethylbenzoic acid, gamma-lactone
PubChem CID 6885
ChEBI CHEBI:38085
IUPAC Name 3H-2-benzofuran-1-one
SMILES C1C2=CC=CC=C2C(=O)O1

Specifications

Melting Point 72°C to 75°C
Density 1.1
Boiling Point 289°C to 291°C
Flash Point 152°C (305°F)
Quantity 100 g
Beilstein 114632
Formula Weight 134.13
Percent Purity ≥98%
Chemical Name or Material Phthalide
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-744-0
RTECSNumber TI3520000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only