missing translation for 'onlineSavingsMsg'
Learn More

Phthalide-3-acetic acid, 98+%

Catalog No. AAL0598406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0598406 5 g
AAL0598414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0598406 Supplier Thermo Scientific Chemicals Supplier No. L0598406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4743-58-2 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4743-58-2
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00051729
InChI Key FJWKEFBYCZSVNZ-UHFFFAOYSA-N
Synonym phthalide-3-acetic acid, 3-oxo-1,3-dihydro-2-benzofuran-1-yl acetic acid, 2-3-oxo-1,3-dihydroisobenzofuran-1-yl acetic acid, 3-oxo-1,3-dihydro-isobenzofuran-1-yl-acetic acid, 2-3-oxo-1,3-dihydro-2-benzofuran-1-yl acetic acid, 3-oxo-1h-2-benzofuran-1-yl acetic acid, 2-3-oxo-1h-2-benzofuran-1-yl acetic acid, 2-3-oxohydroisobenzofuranyl acetic acid, acmc-20ajvy, 1,3-dihydro-3-oxo-1-isobenzofuranacetic acid
PubChem CID 589730
IUPAC Name 2-(3-oxo-1H-2-benzofuran-1-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(OC2=O)CC(=O)O

Specifications

Melting Point 148°C to 153°C
Quantity 5 g
Beilstein 156005
Formula Weight 192.17
Percent Purity ≥98%
Chemical Name or Material Phthalide-3-acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only