missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Hydrazide 98.0+%, TCI America™

Catalog No. p-7134570
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P035425G 25 g
P0354500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P035425G Supplier TCI America Supplier No. P035425G
Only null left

Chemical Identifiers

CAS 1445-69-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00006888
InChI Key JNQUJPBFSQETPT-UHFFFAOYNA-N
Synonym 2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
PubChem CID 219401
IUPAC Name 1,4,4a,8a-tetrahydrophthalazine-1,4-dione
SMILES O=C1N=NC(=O)C2C=CC=CC12

Specifications

Color White
Quantity 25 g
Formula Weight 162.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalic Hydrazide
EINECSNumber (5)-3837
RTECSNumber TH8890080
TSCA No