missing translation for 'onlineSavingsMsg'
Learn More

Phthalic anhydride, ACS reagent

Catalog No. AC423320020
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423320020 2 kg Glass bottle
AC423325000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423320020 Supplier Thermo Scientific Chemicals Supplier No. 423320020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
MDL Number MFCD00005918
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
PubChem CID 6811
ChEBI CHEBI:36605
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

Melting Point 129.0°C to 132.0°C
Color White
Boiling Point 284.0°C
Flash Point 152°C
Ignition Residue 0.01% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2 kg
Beilstein 17,469
Fieser 01,882; 17,148
Merck Index 15,7484
Solubility Information Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water
IUPAC Name 2-benzofuran-1,3-dione
Viscosity 6.4 mPa.s (197°C)
Formula Weight 148.12
Percent Purity 99.0 to 100.2%
Grade ACS Reagent
Physical Form Flaky Crystals
Chemical Name or Material Phthalic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
EINECSNumber 201-607-5
RTECSNumber TI3150000
TSCA TSCA

RUO – Research Use Only