missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, ACS, 99.0-100.2%, Spectrum™ Chemical

Catalog No. p-7166024
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612364 25 g Poly Bottle
18612365 500 g Poly Bottle
18612367 2.5 kg Poly Bottle
18612366 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612364 Supplier Spectrum Chemical Mfg Cor Supplier No. P114025GM
Only null left

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
IUPAC Name 1,3-dihydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Melting Point Within 3°C
Ignition Residue 0.0001
Packaging Poly Bottle
Quantity 25 g
Percent Purity 99 to 100.2%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.