missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, 99%, Thermo Scientific™

Catalog No. AC154960100
Change view
Click to view available options
10kg; Plastic drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC154960100 10kg; Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC154960100 Supplier Thermo Scientific Chemicals Supplier No. 154960100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 85-44-9
Boiling Point 284.0°C
Flash Point 152°C
Assay Percent Range 99%
Packaging Plastic drum
Molecular Formula C8H4O3
Quantity 10kg
MDL Number MFCD00005918
Beilstein 17,469
Fieser 01,882; 17,148
Merck Index 15,7484
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
Solubility Information Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
SMILES O=C1OC(=O)C2=CC=CC=C12
IUPAC Name 2-benzofuran-1,3-dione
Molecular Weight (g/mol) 148.12
PubChem CID 6811
Viscosity 6.4 mPa.s (197°C)
ChEBI CHEBI:36605
Formula Weight 148.12
Percent Purity 99%
Chemical Name or Material Phthalic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
EINECSNumber 201-607-5

RUO – Research Use Only