missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, 99%

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol

Supplier:  Thermo Scientific Chemicals 154960010

 View more versions of this product

Catalog No. AC154960010



Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phthalic anhydride
129.0°C to 132.0°C
152°C
Plastic bottle
MFCD00005918
17,469
15,7484
Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water
O=C1OC(=O)C2=CC=CC=C12
148.12
6.4 mPa.s (197°C)
148.12
85-44-9
284.0°C
99%
C8H4O3
1 kg
01,882; 17,148
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
LGRFSURHDFAFJT-UHFFFAOYSA-N
2-benzofuran-1,3-dione
6811
CHEBI:36605
99%

Chemical Identifiers

85-44-9
148.12
LGRFSURHDFAFJT-UHFFFAOYSA-N
6811
O=C1OC(=O)C2=CC=CC=C12
C8H4O3
MFCD00005918
phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
CHEBI:36605
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 201-607-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only