missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Acid 99.0+%, TCI America™

Catalog No. P028725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P028725G 25 g
P0287500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P028725G Supplier TCI America Supplier No. P028725G

Chemical Identifiers

CAS 88-99-3
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002467
InChI Key XNGIFLGASWRNHJ-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
PubChem CID 1017
ChEBI CHEBI:29069
IUPAC Name phthalic acid
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 210°C
Color White
Formula Weight 166.13
Percent Purity ≥99.0% (GC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Phthalic Acid
EINECSNumber (3)-1337
RTECSNumber TH9625000
TSCA Yes