missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, 99%

Catalog No. AAA1445030
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1445030 250 g
AAA144500B 1000 g
AAA144500I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1445030 Supplier Thermo Scientific Chemicals Supplier No. A1445030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-99-3 | C8H6O4 | 166.132 g/mol

Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

Solubility
Soluble in water, alcohol and etherSoluble in alcohol. Slightly soluble in water, methanol and ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 88-99-3
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002467
InChI Key XNGIFLGASWRNHJ-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
PubChem CID 1017
ChEBI CHEBI:29069
IUPAC Name phthalic acid
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 205°C (decomposition)
Density 1.593
Quantity 250 g
Flash Point 168°C (334°F)
Odor Odorless
Beilstein 608199
Merck Index 14,7371
Solubility Information Soluble in water,alcohol and etherSoluble in alcohol; Slightly soluble in water,methanol and ether; Insoluble in chloroform.
Formula Weight 166.13
Percent Purity 99%
Chemical Name or Material Phthalic acid
Show More Show Less
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 201-873-2
RTECSNumber TH9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only