missing translation for 'onlineSavingsMsg'
Learn More

Phthalhydrazide, 99%

CAS: 1445-69-8 | C8H6N2O2 | 162.15 g/mol

$177.76 - $177.76

Chemical Identifiers

CAS 1445-69-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00006888
InChI Key JNQUJPBFSQETPT-UHFFFAOYNA-N
Synonym 2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
PubChem CID 219401
IUPAC Name 2,3-dihydrophthalazine-1,4-dione
SMILES O=C1N=NC(=O)C2C=CC=CC12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131061000
SDS View Documents
Thermo Scientific Chemicals
131061000
100 g Plastic bottle
Each for $177.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1445-69-8
162.15
JNQUJPBFSQETPT-UHFFFAOYNA-N
219401
O=C1N=NC(=O)C2C=CC=CC12
C8H6N2O2
MFCD00006888
2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
2,3-dihydrophthalazine-1,4-dione

Specifications

1445-69-8
White
Authentic
Plastic bottle
MFCD00006888
24, 371
JNQUJPBFSQETPT-UHFFFAOYNA-N
2,3-dihydrophthalazine-1,4-dione
219401
99%
Phthalhydrazide, 99%
>300°C
1% max.
99%
C8H6N2O2
100 g
2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
O=C1N=NC(=O)C2C=CC=CC12
162.15
162.15
Chunks and/or Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-893-4

RTECSNumber : TH8890080

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only