missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalazine 98.0+%, TCI America™
$63.52 - $194.44
Chemical Identifiers
| CAS | 253-52-1 |
|---|---|
| Molecular Formula | C8H6N2 |
| Molecular Weight (g/mol) | 130.15 |
| MDL Number | MFCD00006908 |
| InChI Key | LFSXCDWNBUNEEM-UHFFFAOYSA-N |
| Synonym | 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine |
| PubChem CID | 9207 |
| ChEBI | CHEBI:36597 |
| IUPAC Name | phthalazine |
| SMILES | C1=CC=C2C=NN=CC2=C1 |
Chemical Identifiers
| 253-52-1 | |
| 130.15 | |
| LFSXCDWNBUNEEM-UHFFFAOYSA-N | |
| 9207 | |
| phthalazine |
| C8H6N2 | |
| MFCD00006908 | |
| 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine | |
| CHEBI:36597 | |
| C1=CC=C2C=NN=CC2=C1 |
Specifications
| 253-52-1 | |
| White-Yellow | |
| C8H6N2 | |
| 5 g | |
| LFSXCDWNBUNEEM-UHFFFAOYSA-N | |
| phthalazine | |
| 9207 | |
| 130.15 | |
| Crystalline Powder |
| 93°C | |
| 317°C | |
| MFCD00006908 | |
| 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine | |
| C1=CC=C2C=NN=CC2=C1 | |
| 130.15 | |
| CHEBI:36597 | |
| ≥98.0% (GC,T) | |
| Phthalazine |
Safety and Handling
TSCA : Yes