missing translation for 'onlineSavingsMsg'
Learn More

Phthalazine 98.0+%, TCI America™

Catalog No. P06945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06945G 5 g
P069425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06945G Supplier TCI America Supplier No. P06945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 253-52-1
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006908
InChI Key LFSXCDWNBUNEEM-UHFFFAOYSA-N
Synonym 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine
PubChem CID 9207
ChEBI CHEBI:36597
IUPAC Name phthalazine
SMILES C1=CC=C2C=NN=CC2=C1

Specifications

Melting Point 93°C
Color White-Yellow
Boiling Point 317°C
Quantity 5 g
Formula Weight 130.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalazine