missing translation for 'onlineSavingsMsg'
Learn More

Phthalazine, 98%

CAS: 253-52-1 | C8H6N2 | 130.15 g/mol

Supplier:  Thermo Scientific Chemicals A1227014

 View more versions of this product

Catalog No. AAA1227014


Only null left
Explore available promotions

Description

Description

Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.

Solubility
Soluble in water, methanol, chloroform.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Phthalazine
89°C to 92°C
C8H6N2
25 g
14,7370
Soluble in water,methanol,chloroform.
C1=CC=C2C=NN=CC2=C1
130.15
CHEBI:36597
98%
253-52-1
317°C
MFCD00006908
109240
2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine
LFSXCDWNBUNEEM-UHFFFAOYSA-N
phthalazine
9207
130.15

Chemical Identifiers

253-52-1
130.15
LFSXCDWNBUNEEM-UHFFFAOYSA-N
9207
phthalazine
C8H6N2
MFCD00006908
2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine
CHEBI:36597
C1=CC=C2C=NN=CC2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 205-963-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only