Learn More
Phthalazine, 98%
CAS: 253-52-1 | C8H6N2 | 130.15 g/mol
Supplier: Thermo Scientific Chemicals A1227006
Description
Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.
Solubility
Soluble in water, methanol, chloroform.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications
| Phthalazine | |
| 89°C to 92°C | |
| C8H6N2 | |
| 5 g | |
| 14,7370 | |
| Soluble in water,methanol,chloroform. | |
| C1=CC=C2C=NN=CC2=C1 | |
| 130.15 | |
| CHEBI:36597 | |
| 98% |
| 253-52-1 | |
| 317°C | |
| MFCD00006908 | |
| 109240 | |
| 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine | |
| LFSXCDWNBUNEEM-UHFFFAOYSA-N | |
| phthalazine | |
| 9207 | |
| 130.15 |
Chemical Identifiers
| 253-52-1 | |
| 130.15 | |
| LFSXCDWNBUNEEM-UHFFFAOYSA-N | |
| 9207 | |
| phthalazine |
| C8H6N2 | |
| MFCD00006908 | |
| 2,3-benzodiazine, benzo d pyridazine, 2,3-diazanaphthalene, beta-phenodiazine, .beta.-phenodiazine, unii-91y28dm24n, ccris 6894, phthalazin, benzo-orthodiazine | |
| CHEBI:36597 | |
| C1=CC=C2C=NN=CC2=C1 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-963-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only