missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalamide 98.0+%, TCI America™
$65.24 - $685.97
Chemical Identifiers
| CAS | 88-96-0 |
|---|---|
| Molecular Formula | C8H8N2O2 |
| Molecular Weight (g/mol) | 164.164 |
| MDL Number | MFCD00025478 |
| InChI Key | NAYYNDKKHOIIOD-UHFFFAOYSA-N |
| Synonym | phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs |
| PubChem CID | 6956 |
| ChEBI | CHEBI:38799 |
| IUPAC Name | benzene-1,2-dicarboxamide |
| SMILES | C1=CC=C(C(=C1)C(=O)N)C(=O)N |
Chemical Identifiers
| 88-96-0 | |
| 164.164 | |
| NAYYNDKKHOIIOD-UHFFFAOYSA-N | |
| 6956 | |
| benzene-1,2-dicarboxamide |
| C8H8N2O2 | |
| MFCD00025478 | |
| phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs | |
| CHEBI:38799 | |
| C1=CC=C(C(=C1)C(=O)N)C(=O)N |
Specifications
| 88-96-0 | |
| White-Yellow | |
| MFCD00025478 | |
| phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs | |
| C1=CC=C(C(=C1)C(=O)N)C(=O)N | |
| 164.164 | |
| CHEBI:38799 | |
| ≥98.0% (N) | |
| Phthalamide |
| 223°C | |
| C8H8N2O2 | |
| 25 g | |
| NAYYNDKKHOIIOD-UHFFFAOYSA-N | |
| benzene-1,2-dicarboxamide | |
| 6956 | |
| 164.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CZ2200000
TSCA : Yes