missing translation for 'onlineSavingsMsg'
Learn More

Phthalamide 98.0+%, TCI America™

Catalog No. P028325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P028325G 25 g
P0283500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P028325G Supplier TCI America Supplier No. P028325G

Chemical Identifiers

CAS 88-96-0
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00025478
InChI Key NAYYNDKKHOIIOD-UHFFFAOYSA-N
Synonym phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs
PubChem CID 6956
ChEBI CHEBI:38799
IUPAC Name benzene-1,2-dicarboxamide
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)N

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Phthalamide
RTECSNumber CZ2200000
TSCA Yes