missing translation for 'onlineSavingsMsg'
Learn More

Phthalamide, 97%

Catalog No. AAL0860009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0860009 10 g
AAL0860018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0860009 Supplier Thermo Scientific Chemicals Supplier No. L0860009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88-96-0 | C8H8N2O2 | 164.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-96-0
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00025478
InChI Key NAYYNDKKHOIIOD-UHFFFAOYSA-N
Synonym phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs
PubChem CID 6956
ChEBI CHEBI:38799
IUPAC Name benzene-1,2-dicarboxamide
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)N

Specifications

Melting Point 223°C (decomposition)
Quantity 10 g
Beilstein 1868220
Merck Index 14,7369
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material Phthalamide
EINECSNumber 201-870-6
RTECSNumber CZ2200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only