missing translation for 'onlineSavingsMsg'
Learn More

Phthalaldehydic Acid 98.0+%, TCI America™

Catalog No. P0281500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0281500G 500 g
P028125G 25 g
P0281100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0281500G Supplier TCI America Supplier No. P0281500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-67-5
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00003336
InChI Key DYNFCHNNOHNJFG-UHFFFAOYSA-N
Synonym 2-carboxybenzaldehyde, phthalaldehydic acid, o-formylbenzoic acid, benzoic acid, 2-formyl, o-carboxybenzaldehyde, benzoic acid, formyl, benzaldehyde-2-carboxylic acid, formylbenzoic acid, carboxybenzaldehyde, pubchem7365
PubChem CID 8406
ChEBI CHEBI:17605
IUPAC Name 2-formylbenzoic acid
SMILES C1=CC=C(C(=C1)C=O)C(=O)O

Specifications

Melting Point 98°C
Color Red-White
Quantity 500 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalaldehydic Acid
EINECSNumber (3)-1670
RTECSNumber TH7015000
TSCA Yes