missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Oakton™ Testr™ Meter Accessories

Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.

Supplier:  Cole-Parmer™ 3565009

 View more versions of this product

Catalog No. 13-200-228


Only null left
Explore available promotions

Description

Description

Handy replacement and accessory items
Specifications

Specifications

1 in.
Double-juntion pH Electrode
ORP Sensor
5.1 in.
ABS
-999 to 1000mV
0°C to 80°C
25.4 mm
Measuring ion activity in water
Double
130 mm
Double
32°F to 176°F
Oakton ORPTestr 10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions