missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Photosynthesis Gas Measurement Apparatus

Catalog No. S15017
Change view
Click to view available options
Grade:
7 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15017 7 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15017 Supplier Eisco™ Supplier No. BI0228

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Photosynthesis Gas Measurement Apparatus allows for the collection and measurement of gas created by aquatic plants during photosynthesis.

  • Gas evolved can be measured by the calibrated capillary tube and drawn off for analysis by the plastic syringes
  • Each vessel has a 1 in. diameter
  • Apparatus measures approximately 6 x 7.5 in.
  • Stand is approximately 4.5 x 8.5 x 4.7 in.
  • Mounted on a plastic panel

Specifications

Product Type Apparatus
Material Plastic
For Use With (Application) Gas Measurement
Includes Capillary Tubes, Syringes, and Base
Dimensions (L x W x H) 4.5 x 8.5 x 4.7 in. (Stand)
Grade 7 to Undergraduate