missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic acid hydrate

Catalog No. AA4011614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4011614 25 g
AA4011622 100 g
AA4011636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4011614 Supplier Thermo Scientific Chemicals Supplier No. 04011614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12067-99-1 | P2O5·24WO3·xH2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphotungstic acid is used as a catalyst for the hydrolysis of propene to prepare 2-propanol. It is also involved in heterogeneous catalysis reaction to prepare propene from 2-propanol and methanol from hydrocarbons. Its ethanolic solution is used in biology as well as to precipitate proteins. It is utilized to precipitate different types of dye such as basic dyes and triphenylmethane dyes. In histology, it is used for staining of cell specimens. It is used in the gravimetric analysis of potassium in analytical chemistry.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong bases, strong oxidizing agents.

Chemical Identifiers

CAS 12067-99-1
MDL Number MFCD00041964
Synonym Tungstophosphoric acid

Specifications

Appearance White to off-white powder
Color White
Density 2.852g/cm3 at 20°C
Odor Odorless
Molecular Formula P2O5·24WO3·xH2O
Quantity 25 g
Physical Form Powder
Solubility Information Soluble in water,alcohol and ether
Formula Weight 5706.35 (Anhydrous)
Chemical Name or Material Phosphotungstic acid hydrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 235-087-6
RTECSNumber TH5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only