missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphotungstic Acid Hydrate, MP Biomedicals™
Phosphotungstic Acid Hydrate is used for cell staining.
$123.63 - $123.63
Chemical Identifiers
| CAS | 12501-23-4 |
|---|---|
| Molecular Formula | H5O41PW12 |
| Molecular Weight (g/mol) | 2880.20 |
| MDL Number | MFCD00011342 |
| InChI Key | AVFBYUADVDVJQL-UHFFFAOYSA-N |
| Synonym | phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade |
| PubChem CID | 16212977 |
| IUPAC Name | 12-Tungstophosphate hydrate |
| SMILES | O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP215624325
|
MP Biomedicals Inc
0215624325 |
25 g |
Each for $123.63
|
|
|||||
Description
- Phosphotungstic acid is used in histology as a component for staining of cell specimens.
- It is a common negative stain for viruses, nerves, polysaccharides, and other biological tissue materials for imaging by a transmission electron microscope.
- Phosphotungstic acid has been used to precipitate different types of dyes as lakes.
Chemical Identifiers
| 12501-23-4 | |
| 2880.20 | |
| AVFBYUADVDVJQL-UHFFFAOYSA-N | |
| 16212977 | |
| O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O |
| H5O41PW12 | |
| MFCD00011342 | |
| phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade | |
| 12-Tungstophosphate hydrate |
Specifications
| White | |
| 12501-23-4 | |
| H5O41PW12 | |
| MFCD00011342 | |
| Soluble in water (1 g/10 mL), alcohol and ether. | |
| O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O | |
| 2880.20 | |
| 2880.2 (anhyd) |
| ∼95°C | |
| 25 g | |
| Crystalline Powder | |
| phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade | |
| AVFBYUADVDVJQL-UHFFFAOYSA-N | |
| 12-Tungstophosphate hydrate | |
| 16212977 |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C).
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.