missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydre d’acide phosphotungstique (cristallin/certifié) ACS, Fisher Chemical™
Spécifications
Spécifications
| Nom ou substance chimique | Phosphotungstic Acid |
| Couleur | Blanc |
| Point de fusion | 89 °C |
| Métaux lourds (sous forme de Pb) | 0,005% max. |
| Désignation | Test Réussi |
| Emballage | Bouteille en polyéthylène |
| Quantité | 25 g |
| Forme physique | Solide |
| Numéro UN | 3260 |
| Niveau | Certifié |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. . NFPA Health:3 Flammability:0 Instability:0 |
| DOTInformation | DOT Class 8, : Corrosive |
| Recommended Storage | Réfrigérée |