missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Phosphorus(V) Oxychloride

Catalog No. 201170250G
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
201170250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 201170250G Supplier Sigma Aldrich Supplier No. 201170250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphorus(V) Oxychloride

Specifications

CAS 10025-87-3
Melting Point 1.25°C
Density 1.645 g/mL at 25°C
Boiling Point 105.8°C
Molecular Formula Cl3OP
Linear Formula POCl3
Quantity 250 g
MDL Number MFCD00011443
Synonym Phosphorus(V) oxide chloride; Phosphoryl chloride
Molecular Weight (g/mol) 153.33 g/mol