missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxide, ACS, 98% min

Catalog No. p-4824869
Change view
Click to view available options
Quantity:
50 g
500 g
4 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1052418 50 g
AA1052436 500 g
AA1052457 4 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1052418 Supplier Thermo Scientific Chemicals Supplier No. 01052418
Only null left

For Use As a Drying and Dehydrating Agent, As a Condensing Agent in organic Synthesis, in Sugar Refining | CAS: 1314-56-3 | O5P2 | 141.94 g/mol

Used as a drying and dehydrating agent, condensation reagent in organic synthesis and a laboratory reagent. Used in sugar refining, and in fire extinguishing. Phosphorus pentoxide in DMSO forms Onodera reagent, which oxidizes alcohols. Capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 98% min.
Synonym Phosphoric anhydride, Phosphoric oxide
Formula Weight 141.94
Grade ACS
Odor Odorless
Appearance White powder
Chemical Name or Material Phosphorus(V) oxide
Hazard Category H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 215-236-1

RUO – Research Use Only