missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus(V) oxide, 99.99%
Description
Phosphorus(V) oxide is used as a drying and dehydrating agent, a condensation reagent in organic synthesis and a laboratory reagent. It is also used in sugar refining and in fire extinguishing. Phosphorus pentoxide in DMSO forms an Onodera reagent which oxidizes alcohols. It is capable of converting mineral acids to anhydrides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 360°C (sublimation) |
| Melting Point | 340°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.99% |
| Solubility Information | Soluble in sulfuric acid. Insoluble in acetone and ammonia. Decomposes in water. |
| Formula Weight | 141.94 |
| Odor | Odorless |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | DOT Class: 8, Packaging Group: II |
| EINECSNumber | 215-236-1 |
| RTECSNumber | TH3945000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only