missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4782050
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1184914 25 g
AA1184922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1184914 Supplier Thermo Scientific Chemicals Supplier No. 01184914
Only null left

Phosphorus(V) chloride is used in organic synthesis, chemical reagents and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-13-8
Molecular Formula Cl5P
Molecular Weight (g/mol) 208.224
MDL Number MFCD00011439
InChI Key UHZYTMXLRWXGPK-UHFFFAOYSA-N
Synonym phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
PubChem CID 24819
ChEBI CHEBI:30335
IUPAC Name pentachloro-$l^{5}-phosphane
SMILES P(Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 162° (sublimation)
Color Yellow
Physical Form Solid
Quantity 25 g
Assay Percent Range 99.998% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 208.24
Odor Pungent
Sensitivity Moisture sensitive
Appearance Yellowish crystals
Vapor Pressure 0.02 hPa at 20°C
Density 2.114 g/cm3 at 20°C
Chemical Name or Material Phosphorus(V) chloride
Show More Show Less
Hazard Category H302-H314-H330-H373
Hazard Statement Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 233-060-3
RTECSNumber TB6125000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only