missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals

For Use As Dehydrating and Chlorinating Agent, Catalyst

Supplier:  Thermo Scientific Chemicals 01052336

 View more versions of this product

Catalog No. AA1052336

Only null left
Explore available promotions

Description

Description

Phosphorus(V) chloride can be used for both oxidative and substitutive chlorinations, & for both organic and inorganic compounds. Dehydrating agent. Precursor to Vilsmeier reagent, lithium hexafluorophosphate electrolyite for lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phosphorus(V) chloride
10026-13-8
Solid
98%
MFCD00011439
Soluble in carbon disulfide and carbon tetrachloride.
P(Cl)(Cl)(Cl)(Cl)Cl
208.224
CHEBI:30335
Pungent
Yellowish crystals
2.114 g/cm3 at 20°C
162° (sublimation)
Yellow
500 g
Cl5P
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
UHZYTMXLRWXGPK-UHFFFAOYSA-N
pentachloro-$l^{5}-phosphane
24819
208.24
Moisture sensitive
0.02 hPa at 20°C

Chemical Identifiers

10026-13-8
208.224
UHZYTMXLRWXGPK-UHFFFAOYSA-N
24819
pentachloro-$l^{5}-phosphane
Cl5P
MFCD00011439
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
CHEBI:30335
P(Cl)(Cl)(Cl)(Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H330-H373

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 233-060-3

RTECSNumber : TB6125000

TSCA : Yes

storageNote1 : Moisture Sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only