missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) bromide, 95%

Catalog No. AAB2248136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2248136 500 g
AAB2248114 25 g
AAB2248122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2248136 Supplier Thermo Scientific Chemicals Supplier No. B2248136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7789-69-7 | Br5P | 430.494 g/mol

Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 7789-69-7
Molecular Formula Br5P
Molecular Weight (g/mol) 430.494
MDL Number MFCD00011437
InChI Key QRKVRHZNLKTPGF-UHFFFAOYSA-N
Synonym phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromide
PubChem CID 62678
IUPAC Name pentabromo-$l^{5}-phosphane
SMILES P(Br)(Br)(Br)(Br)Br

Specifications

Boiling Point 105°C to 107°C (decomposition)
Melting Point ∽105°C (decomposition)
Quantity 500 g
UN Number UN2691
Beilstein 3903332
Merck Index 14,7350
Solubility Information Reacts with water.
Formula Weight 430.5
Percent Purity 95%
Odor Acrid
Sensitivity Moisture sensitive
Chemical Name or Material Phosphorus(V) bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTABROMIDE
EINECSNumber 232-186-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only