missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus tribromide, 99%

Catalog No. AC169475000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169475000 500 g Glass bottle
AC169471000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169475000 Supplier Thermo Scientific Chemicals Supplier No. 169475000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-60-8 | Br3P | 270.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7789-60-8
Molecular Formula Br3P
Molecular Weight (g/mol) 270.69
MDL Number MFCD00011436
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
PubChem CID 24614
IUPAC Name tribromophosphane
SMILES P(Br)(Br)Br

Specifications

Boiling Point 175.0°C
Melting Point -40.0°C
Color Brown or Colorless to Yellow
Physical Form Liquid
Quantity 500 g
Assay Percent Range 98.5% min. (ex Br) (Argentometry)
Linear Formula PBr3
Fieser 01,873; 02,330; 07,292
Merck Index 15, 7469
Solubility Information Solubility in water: reacts. Other solubilities: soluble in acetone, alcohol, carbon disulfide,, hydrogen sulfide, methanol, ether, chloroform and, carbon tetrachloride
Formula Weight 270.69
Percent Purity 99%
Packaging Glass bottle
Refractive Index 1.697
Specific Gravity 2.88
Density 2.8800g/mL
Chemical Name or Material Phosphorus tribromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 232-178-2

RUO – Research Use Only