missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus tribromide, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog No. p-3213053
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC382061000 100 mL AcroSeal™ Glass bottle
AC382068000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC382061000 Supplier Thermo Scientific Chemicals Supplier No. 382061000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Specifications

Chemical Name or Material Phosphorus tribromide
Name Note 1.0M Solution in Dichloromethane
Color Colorless to Yellow
CAS 7789-60-8, 75-09-2
Physical Form Liquid
Quantity 100 mL
Molecular Formula Br3P
Linear Formula PBr2
MDL Number MFCD00011436
Merck Index 15, 7469
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
SMILES P(Br)(Br)Br
IUPAC Name tribromophosphane
Molecular Weight (g/mol) 270.69
PubChem CID 24614
Formula Weight 270.69
Concentration or Composition (by Analyte or Components) 0.9 to 1.1M
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.488
Density 1.4880g/mL
Show More Show Less

RUO – Research Use Only