missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Pentoxide, ACS, 98%, Spectrum™ Chemical

Catalog No. 18612357
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612357 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18612357 Supplier Spectrum Chemical Mfg Cor Supplier No. P1125500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1314-56-3
CAS Max % 1
Quantity 500 g
Percent Purity 0.98
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.