missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus pentoxide, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 340°C |
| CAS | 1314-56-3 |
| Quantity | 50 g |
| Linear Formula | P2O5 |
| Formula Weight | 141.94 |
| Percent Purity | ≥99.99% (trace metals basis) |
| Packaging | Poly Bottle |
| Vapor Pressure | 10 mmHg (238°C) |
| Density | 2.3 g/mL (at 25°C); Vapor Density: 4.9 (vs air) |
Safety and Handling
| RTECSNumber | TH3945000 |