missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus pentoxide, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. 111019227
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019227 50 g
111018231 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019227 Supplier Merck Emd Millipore Supplier No. 43141950G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphorus pentoxide (P2O5) is a commonly used drying agent. It is a component of Eatons reagent, which is commonly used as a catalyst for dehydration reactions.

Chemical Identifiers

Synonym Phosphoric anhydride; Phosphorus(V) oxide

Specifications

Melting Point 340°C
CAS 1314-56-3
Quantity 50 g
Linear Formula P2O5
Formula Weight 141.94
Percent Purity ≥99.99% (trace metals basis)
Packaging Poly Bottle
Vapor Pressure 10 mmHg (238°C)
Density 2.3 g/mL (at 25°C); Vapor Density: 4.9 (vs air)
RTECSNumber TH3945000