missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus pentabromide, 95%

Catalog No. AC428271000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428271000 100 g Glass bottle
AC428270250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428271000 Supplier Thermo Scientific Chemicals Supplier No. 428271000

CAS: 7789-69-7 | Br5P | 430.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-69-7
Molecular Formula Br5P
Molecular Weight (g/mol) 430.49
MDL Number MFCD00011437
InChI Key QRKVRHZNLKTPGF-UHFFFAOYSA-N
Synonym phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromide
PubChem CID 62678
IUPAC Name pentabromo-$l^{5}-phosphane
SMILES P(Br)(Br)(Br)(Br)Br

Specifications

Melting Point 105°C to 107°C
Quantity 100 g
Assay Percent Range 94% min. (Argentometry)
Linear Formula PBr5
Merck Index 15,7462
Solubility Information Solubility in water: reacts
Formula Weight 430.49
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Phosphorus pentabromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-186-6

RUO – Research Use Only