missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Oxychloride, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P11151KG

 View more versions of this product

Catalog No. 18-612-352


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10025-87-3
1 kg
Reagent
1
98%
Steel Can

Chemical Identifiers

10025-87-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.