missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(III) fluoride, 99%

Catalog No. AA3952518
Change view
Click to view available options
Quantity:
50 g
Packaging:
Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA3952518 50 g Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3952518 Supplier Thermo Scientific Chemicals Supplier No. 03952518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-55-3 | F3P | 87.969 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-55-3
Molecular Formula F3P
Molecular Weight (g/mol) 87.969
MDL Number MFCD00040325
InChI Key WKFBZNUBXWCCHG-UHFFFAOYSA-N
Synonym phosphorus trifluoride, trifluorophosphine, phosphorous fluoride, phosphorous trifluoride, phosphorus fluoride, trifluorophosphin, unii-496073dybf, phosphorus iii fluoride, pf3, trifluoridophosphorus
PubChem CID 62665
ChEBI CHEBI:30205
IUPAC Name trifluorophosphane
SMILES FP(F)F

Specifications

Boiling Point -101.5°C
Melting Point -151.5°C
Color Colorless
Physical Form Gas
Quantity 50 g
UN Number UN1955
Merck Index 14,7359
Formula Weight 87.97
Percent Purity 99%
Odor Odorless
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.907 g/mL
Chemical Name or Material Phosphorus(III) fluoride
Show More Show Less
Hazard Category H302+H312-H314-H330-H335
Hazard Statement GHS H Statement
H330-H314-H318-H302-H312
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 2
EINECSNumber 232-008-7
RTECSNumber TH3850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only