missing translation for 'onlineSavingsMsg'
Learn More

Phosphorous Acid, Spectrum™ Chemical

$18575.20 - $18575.20

Chemical Identifiers

CAS 13598-36-2
Molecular Weight (g/mol) 82
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18-612-349
View Documents
Spectrum Chemical Mfg Cor
P110025KGBL
25 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13598-36-2
82

Specifications

White Crystallin
1
25 kg
≥98.5%
Phosphorous Acid
13598-36-2
Amber Glass Bottle
82
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.