missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥85%, Honeywell Fluka™

Catalog No. 6001133
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001133 1 L
6001134 2.5 L
60045821 6 x 1 L
60045820 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001133 Supplier Honeywell Chemicals Supplier No. 304171L
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥85%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Density 1.685g/mL at 25°C
Vapor Pressure 2.2mmHg (20°C), 5mmHg (25°C)
Quantity 1 L
Linear Formula H3PO4
UN Number 1805
Beilstein 1921286
Formula Weight 98g/mol
Percent Purity ≥85%
Grade ACS/puriss. p.a.
Chemical Name or Material Phosphoric Acid, Reag. Ph. Eur. reag. ISO
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314
EINECSNumber 231-633-2