missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Honeywell Fluka™

Catalog No. 60044535
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044535 6 x 1 L
6001140 1 L
6001141 2.5 L
6001142 5 L
60044533 4 x 2.5 L
60044534 4 x 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60044535 Supplier Honeywell Chemicals Supplier No. 041076X1L
Only null left

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., 85.0-88.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Density 1.685g/mL at 25°C
Vapor Pressure 5mmHg (25°C), 2.2mmHg (20°C)
Quantity 6 x 1 L
Linear Formula H3PO4
UN Number UN1805
Formula Weight 98
Percent Purity 85.0 to 88.0%
Grade puriss.
Chemical Name or Material Phosphoric Acid, Meets analytical specification of Ph. Eur.BP, NF, FCC
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H290-H314
DOTInformation 8 / PGIII
EINECSNumber 231-633-2