missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Honeywell Fluka™

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., 85.0-88.0%

Supplier:  Honeywell Chemicals 041074X25L

 View more versions of this product

Catalog No. 60044533

Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Phosphoric Acid
7664-38-2
5mmHg (25°C), 2.2mmHg (20°C)
H3O4P
MFCD00011340
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
OP(=O)(O)O
97.994
CHEBI:26078
85.0 to 88.0%
Meets analytical specification of Ph. Eur.BP, NF, FCC
1.685g/mL at 25°C
4 x 2.5 L
H3PO4
UN1805
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
1004
98
puriss.

Chemical Identifiers

7664-38-2
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid
H3O4P
MFCD00011340
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338-P310

H290-H314

DOTInformation : 8 / PGIII

EINECSNumber : 231-633-2

SDS
Documents

Documents

Promotions

Promotions