missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, Purified, 85% (w/w), Reagents
Phosphoric Acid, Purified, 85% (w/w), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
$160.61 - $3138.11
Chemical Identifiers
| CAS | 7664-38-2 |
|---|---|
| Molecular Formula | H3O4P |
| Molecular Weight (g/mol) | 97.99 |
| InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
| Synonym | Orthophosphoric Acid, o-Phosphoric Acid |
| IUPAC Name | phosphoric acid |
| SMILES | OP(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-816
|
Labchem Inc
C510200015KG |
15 kg |
Each for $1,640.46
|
|
|||||
|
01-336-815
|
Labchem Inc
C510200025D1 |
2.5 L |
Each for $500.71
|
|
|||||
|
01-336-814
|
Labchem Inc
C510200020E7 |
20 L |
Each for $3,138.11
|
|
|||||
|
01-336-813
|
Labchem Inc
C5102000500C |
500 mL |
Each for $160.61
|
|
|||||
Chemical Identifiers
| 7664-38-2 | |
| 97.99 | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| OP(O)(O)=O |
| H3O4P | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid |
Specifications
| <2 | |
| 7664-38-2,7732-18-5 | |
| Cubitainer | |
| 15 kg | |
| Liquid | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| Pure |
| Colorless | |
| 260°C | |
| 1.69 | |
| H3O4P | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| OP(O)(O)=O | |
| 97.99 | |
| Phosphoric Acid, Purified, 85% (w/w) |