missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, NF, 85-88%, Spectrum™ Chemical
PH175, 7664-38-2, H3PO4
Supplier: Spectrum Chemical Mfg Cor PH17525LTPV
Description
Spectrum™ Chemical Phosphoric Acid, NF - All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| Phosphoric Acid | |
| 85 to 88%,12 to 15% | |
| 2.5 L | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| 85 to 88% |
| 7664-38-2,7732-18-5 | |
| Poly-Coated Glass Bottle | |
| H3O4P | |
| OP(O)(O)=O | |
| 97.99 | |
| NF |
Chemical Identifiers
| 7664-38-2 | |
| 97.99 | |
| phosphoric acid |
| H3O4P | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| OP(O)(O)=O |